Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TFG Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody has been used in 2 publications

$206.50 - $426.00

Specifications

Antigen TFG
Concentration 0.5 mg/mL
Dilution Western Blot 0.5-2 ug/ml, Immunohistochemistry, Immunocytochemistry/Immunofluorescence reported in scientific literature (PMID 29093022), Immunohistochemistry-Paraffin 10 ug/ml
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP224485SS
SDP
View Documents
Novus Biologicals
NBP224485SS
0.025 mg
Each for $206.50
Only null left
Add to Cart
 
NBP224485
SDP
View Documents
Novus Biologicals
NBP224485
0.1 mg
Each for $426.00
Only null left
Add to Cart
 
Description

Description

TFG Polyclonal specifically detects TFG in Human, Rat, Primate samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications

Specifications

TFG
Western Blot 0.5-2 ug/ml, Immunohistochemistry, Immunocytochemistry/Immunofluorescence reported in scientific literature (PMID 29093022), Immunohistochemistry-Paraffin 10 ug/ml
Polyclonal
Rabbit
Signal Transduction
0.2 ml PBS and 0.05% BSA with 0.05% Sodium Azide
10342
A portion of amino acids 350-400 of human TFG was used as the immunogen for this antibody.
Primary
Store at -20C. Avoid freeze-thaw cycles.
0.5 mg/mL
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Unconjugated
RUO
Human, Rat, Primate
FLJ36137, protein TFG, TF6, TRK-fused gene, TRK-fused gene protein, TRKT3, TRKT3 oncogene
TFG
IgG
Affinity Purified
The USP13 amino acid sequence used as immunogen for the antibody is 100% homologous in human, chimpanzee, rhesus monkey, horse, mouse, cow and dog, 94% homologous in rat, 88% homologous in opossum and 82% homologous in chicken.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.