Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TFG Antibody (TFG-03), DyLight 405, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP262212V
Description
TFG Monoclonal specifically detects TFG in Human samples. It is validated for Western Blot, Immunocytochemistry.Specifications
TFG | |
Monoclonal | |
DyLight 405 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
TFG | |
Human recombinant protein Trx-his-NTFG | |
0.1 mL | |
Primary | |
The antibody MEM-57 reacts with gamma-epsilon and delta-epsilon dimers of human CD3 complex, a part of a bigger multisubunit T cell receptor complex (CD3/TCR) expressed on peripheral blood T lymphocytes and mature thymocytes. | |
Store at 4°C in the dark. |
Western Blot, Immunocytochemistry | |
TFG-03 | |
Western Blot, Immunocytochemistry | |
FLJ36137, protein TFG, TF6, TRK-fused gene, TRK-fused gene protein, TRKT3, TRKT3 oncogene | |
Mouse | |
Protein A purified | |
Signal Transduction | |
10342 | |
Human | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction