Learn More
TFG Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A302341AT
Description
The recommended shelf life for this product is 1 year from date of receipt.
TFG protein, a recently identified novel protein, is a human homolog of mouse TFG protein. It contains an N-terminal coiled-coil structure, a SPYGQ-rich region followed by octicosapeptide repeats. Research shows that TFG can modulate SHP-1 activity and is a novel member of the NF-kB pathway. The physiological functions of this protein remain largely elusive, but TFG is thought to function by associating with other proteins, including TANK and NEMO. TFG was first identified as a partner of NTRK1 and is involved in generating the thyroid TRK-T3 oncogene, as well as oncogenic rearrangements with ALK in anaplastic lymphoma and NOR1 in mixoid chondrosarcoma.Specifications
| TFG | |
| Polyclonal | |
| TBS with 0.1% BSA and 0.09% sodium azide | |
| Q92734 | |
| TFG | |
| Between 125 and 175 | |
| 20 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
| Western Blot | |
| Unconjugated | |
| TFG | |
| TFG | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 10342, 21787 | |
| 4° C | |
| Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.