Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TFIIB Antibody, HRP, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP149982H
Description
TFIIB Polyclonal antibody specifically detects TFIIB in Human samples. It is validated for Western BlotSpecifications
TFIIB | |
Polyclonal | |
Western Blot | |
general transcription factor IIB, General transcription factor TFIIB, S300-II, TFIIBTF2BRNA polymerase II transcription factor IIB, transcription initiation factor IIB | |
The immunogen for this product maps to a region between residue 256 and 316 of human General Transcription Factor IIB using the numbering given in entry NP_001505.1 (GeneID 2959). | |
0.1 mL | |
DNA replication Transcription Translation and Splicing | |
2959 | |
Store at 4°C in the dark. | |
IgG |
Western Blot | |
HRP | |
PBS | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction