Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TFIIB Antibody (OTI1H10), DyLight 550, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP274492R
Description
TFIIB Monoclonal specifically detects TFIIB in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
TFIIB | |
Monoclonal | |
DyLight 550 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
GTF2B | |
Recombinant protein fragment corresponding to amino acids 43-316 of human TFIIB (NP_001505) produced in E.coli. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat | |
IgG1 |
Western Blot | |
OTI1H10 | |
Western Blot | |
general transcription factor IIB, General transcription factor TFIIB, S300-II, TFIIBTF2BRNA polymerase II transcription factor IIB, transcription initiation factor IIB | |
Mouse | |
Affinity Purified | |
DNA replication Transcription Translation and Splicing | |
2959.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction