Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TFIIB Antibody (SR1524), Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP32220750UL
Description
TFIIB Monoclonal antibody specifically detects TFIIB in Human, Mouse, Rat samples. It is validated for Western Blot, ImmunoprecipitationSpecifications
TFIIB | |
Monoclonal | |
Unconjugated | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Western Blot, Immunoprecipitation | |
SR1524 | |
Western Blot 1:500-1:2000, Immunoprecipitation 1:50 | |
general transcription factor IIB, General transcription factor TFIIB, S300-II, TFIIBTF2BRNA polymerase II transcription factor IIB, transcription initiation factor IIB | |
A synthesized peptide derived from human TFIIB (Uniprot #: Q00403) | |
50 μg | |
DNA replication Transcription Translation and Splicing | |
2959 | |
Store at -20° C. Avoid freeze/thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction