Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TFIIE beta Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP220610
Description
TFIIE beta Polyclonal specifically detects TFIIE beta in Human samples. It is validated for Western Blot, Chromatin Immunoprecipitation, Immunoprecipitation.Specifications
TFIIE beta | |
Polyclonal | |
Unconjugated | |
PBS (pH 7.0), 1.0% BSA and 20% Glycerol with 0.01% Thimerosal | |
FE, General transcription factor IIE subunit 2, general transcription factor IIE, polypeptide 2 (beta subunit, 34kD), general transcription factor IIE, polypeptide 2, beta 34kDa, TF2E2TFIIE-beta, TFIIE-B, transcription initiation factor IIE subunit beta | |
Rabbit | |
33 kDa | |
0.1 mL | |
DNA replication Transcription Translation and Splicing | |
2961 | |
Human | |
IgG |
Western Blot, Immunoprecipitation, ChIP Assay | |
1mg/mL | |
Western Blot 1:1000-1:10000, Immunoprecipitation 1:100-1:500, Chromatin Immunoprecipitation (ChIP) | |
P29084 | |
GTF2E2 | |
Recombinant protein encompassing a sequence within the center region of human TFIIE beta. The exact sequence is proprietary. | |
Affinity Purified | |
RUO | |
Primary | |
Human. Rhesus Monkey (100%). | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction