Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TFIP11 Mouse anti-Human, Clone: 1A1C8, Proteintech

Mouse Monoclonal Antibody
$518.60
Specifications
Antigen | TFIP11 |
---|---|
Clone | 1A1C8 |
Concentration | 0.95 mg/mL |
Applications | Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-6550
![]() ![]() |
Proteintech Group Inc
661141IG150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
This gene encodes a protein component of the spliceosome that promotes the release of the lariat-intron during late-stage splicing through the recruitment of a pre-mRNA splicing factor called DEAH-box helicase 15. The encoded protein contains a G-patch domain, a hallmark of RNA-processing proteins, that binds DEAH-box helicase 15. This protein contains an atypical nuclear localization sequence as well as a nuclear speckle-targeting sequence, enabling it to localize to distinct speckled regions within the cell nucleus. Polymorphisms in this gene are associated with dental caries suggesting a role in amelogenesis. Alternative splicing results in multiple transcript variants.Specifications
TFIP11 | |
0.95 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
bK445C9.6, STIP, STIP 1, TFIP11, TIP39 | |
TFIP11 | |
IgG2b | |
Protein A | |
Antibody |
1A1C8 | |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Mouse | |
Human | |
Q9UBB9 | |
24144 | |
TFIP11 Fusion Protein Ag5821 | |
Primary | |
-20°C | |
TFIP11 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title