Learn More
Invitrogen™ TFIP11 Polyclonal Antibody

Description
Immunogen sequence: MSLSHLYRDG EGRIDDDDDE RENFEITDWD LQNEFNPNRQ RHWQTKEEAT YGVWAERDSD DERPSFGGKR ARDYSAPVNF ISAGLKKGAA EEAELEDSDD EEKPVKQDDF PKDFGPRKLK TGGNFKPSQK GFAGGTKSFM; Positive Samples: LO2, HeLa, A-431, MCF-7, HT-1080, Mouse brain, Mouse liver; Cellular Location: Cytoplasm, Nucleus.
Specifications
Specifications
| Antigen | TFIP11 |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Concentration | 2.31 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | TFIP11 |
| Gene Accession No. | Q9ERA6, Q9UBB9 |
| Gene Alias | 2810002G02Rik; AF097181; AW046167; bK445C9.6; hNtr1; HSPC006; NTR1; Septin and tuftelin-interacting protein 1; Spp382; Srr1; STIP; STIP-1; Tfip11; TIP33; TIP39; tuftelin interacting protein 11; tuftelin-interacting protein 11; tuftelin-interacting protein 33; tuftelin-interacting protein 39; tuftelin-interacting protein, 39 kD |
| Gene Symbols | TFIP11 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.