Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TFIP11 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
SDP

Catalog No. 501729451 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-172-9451 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-172-9451 Supplier Proteintech Group Inc Supplier No. 144361AP150UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This gene encodes a protein component of the spliceosome that promotes the release of the lariat-intron during late-stage splicing through the recruitment of a pre-mRNA splicing factor called DEAH-box helicase 15. The encoded protein contains a G-patch domain, a hallmark of RNA-processing proteins, that binds DEAH-box helicase 15. This protein contains an atypical nuclear localization sequence as well as a nuclear speckle-targeting sequence, enabling it to localize to distinct speckled regions within the cell nucleus. Polymorphisms in this gene are associated with dental caries suggesting a role in amelogenesis. Alternative splicing results in multiple transcript variants.

Specifications

Antigen TFIP11
Applications Immunocytochemistry, Immunofluorescence, Western Blot
Classification Polyclonal
Concentration 0.16 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene TFIP11
Gene Accession No. Q5U2Y6, Q9ERA6, Q9UBB9
Gene Alias bK445C9.6, STIP, STIP 1, TFIP11, TIP39
Gene Symbols Tfip11
Host Species Rabbit
Immunogen TFIP11 Fusion Protein Ag5821
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 24144, 288718, 54723
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.