Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TFPI-2 Antibody (MM0564-9F6) - Azide and BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
$206.50 - $560.50
Specifications
Antigen | TFPI-2 |
---|---|
Clone | MM0564-9F6 |
Concentration | LYOPH |
Applications | Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunohistochemistry, Western Blot |
Classification | Monoclonal |
Description
TFPI-2 Monoclonal specifically detects TFPI-2 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.Specifications
TFPI-2 | |
LYOPH | |
Monoclonal | |
Purified | |
RUO | |
P48307 | |
7980 | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
MM0564-9F6 | |
Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunohistochemistry, Western Blot | |
Unconjugated | |
Mouse | |
Human | |
Placental protein 5, PP5FLJ21164, TFPI-2REF1, tissue factor pathway inhibitor 2 | |
TFPI2 | |
Protein G purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title