Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TFPI-2 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501981375
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-1375 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-1375 Supplier Bioss Supplier No. BS1144R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

TFPI2 may play a role in the regulation of plasmin-mediated matrix remodeling. It inhibits trypsin, plasmin, factor VIIa/tissue factor and weakly factor Xa. TFPI2 has no effect on thrombin.

Specifications

Antigen TFPI-2
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene TFPI2
Gene Accession No. P48307
Gene Alias AV000670; Placental protein 5; PP5; PP5/TFPI-2; PP5FLJ21164; REF1; REF-1; retinal pigment epithelium cell factor 1; Tfpi2; TFPI-2; TFPI-2REF1; Tissue factor pathway inhibitor 2
Gene Symbols TFPI2
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human TFPI2.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7980
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.