Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TFPI Antibody (120), Biotin

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP290441B
Description
TFPI Monoclonal specifically detects TFPI in Mouse samples. It is validated for ELISA.Specifications
TFPI | |
Monoclonal | |
Biotin | |
PBS | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Has cross-reactivity in ELISA with: Human TFPI | |
Store at 4°C in the dark. | |
IgG |
ELISA | |
120 | |
ELISA | |
convertin, EPITFPI1LACITFI, Extrinsic pathway inhibitor, Lipoprotein-associated coagulation inhibitor, tissue factor pathway inhibitor, tissue factor pathway inhibitor (lipoprotein-associated coagulation inhibitor) | |
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse TFPI (NP_035706.1; Met 1-Lys 289). | |
0.1 mL | |
Cardiovascular Biology | |
7035 | |
Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction