Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TFPI Antibody (3A8), Novus Biologicals™

Human Monoclonal Antibody
$302.50 - $494.50
Specifications
Antigen | TFPI |
---|---|
Clone | 3A8 |
Dilution | ELISA |
Applications | ELISA |
Classification | Monoclonal |
Description
TFPI Monoclonal antibody specifically detects TFPI in Human, Rabbit samples. It is validated for ELISASpecifications
TFPI | |
ELISA | |
Monoclonal | |
Purified | |
RUO | |
Human, Rabbit | |
convertin, EPITFPI1LACITFI, Extrinsic pathway inhibitor, Lipoprotein-associated coagulation inhibitor, tissue factor pathway inhibitor, tissue factor pathway inhibitor (lipoprotein-associated coagulation inhibitor) | |
Recombinant Human TFPI protein [UniProt P10646] | |
Primary | |
Store at -20 to -70C. Avoid freeze-thaw cycles. |
3A8 | |
ELISA | |
Unconjugated | |
Human | |
Cardiovascular Biology | |
50% Glycerol, 0.01M PBS, PH 7.4 | |
7035 | |
IgG1 | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title