Learn More
Invitrogen™ TFPI Polyclonal Antibody

Description
PA1-43059 detects Tissue Factor Pathway Inhibitor in human samples. PA1-43059 has successfully been used in Western blot and ELISA applications. PA1-43059 was derived from human tissue factor pathway inhibitor.
Specifications
Specifications
| Antigen | TFPI |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Concentration | 9.3 mg/mL |
| Conjugate | Unconjugated |
| Formulation | 50% water with 50% glycerol and no preservative |
| Gene | TFPI |
| Gene Accession No. | P10646 |
| Gene Alias | A630013F22Rik; anti-convertin; AW552122; EPI; Extrinsic pathway inhibitor; LACI; Lipoprotein-associated coagulation inhibitor; OTTHUMP00000163472; OTTHUMP00000205431; TFI; TFPI; TFPI1; Tissue factor pathway inhibitor; tissue factor pathway inhibitor (lipoprotein-associated coagulation inhibitor) |
| Gene Symbols | TFPI |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.