Learn More
Invitrogen™ TFPI Polyclonal Antibody

Description
Immunogen sequence: DSEEDEEHTI ITDTELPPLK LMHSFCAFKA DDGPCKAIMK RFFFNIFTRQ CEEFIYGGCE GNQNRFESLE ECKKMCTRDN ANRIIKTTLQ QEKPDFCFLE EDPGICRGYI TRYFYNNQTK QCERFKYGGC LGNMNNFETL EECKNICEDG PNGFQVDNYG TQLNAVNNSL TPQSTKVPSL FVTKEGTNDG WKNAAH; Positive Samples: MCF-7, Mouse spleen, Rat lung; Cellular Location: GPI-anchor, Lipid-anchor, Microsome membrane.
Specifications
Specifications
| Antigen | TFPI |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Concentration | 1.36 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 |
| Gene | TFPI |
| Gene Accession No. | O54819, P10646, Q02445 |
| Gene Alias | A630013F22Rik; anti-convertin; AW552122; EPI; Extrinsic pathway inhibitor; LACI; Lipoprotein-associated coagulation inhibitor; OTTHUMP00000163472; OTTHUMP00000205431; TFI; TFPI; TFPI1; Tissue factor pathway inhibitor; tissue factor pathway inhibitor (lipoprotein-associated coagulation inhibitor) |
| Gene Symbols | TFPI |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.