Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TFPI2 Monoclonal Antibody (375717)
GREENER_CHOICE

Catalog No. PIMA524116
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA524116 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA524116 Supplier Invitrogen™ Supplier No. MA524116
Add to Cart
Add to Cart

Rat Monoclonal Antibody

In direct ELISAs and Western blots, no cross-reactivity with recombinant mouse TFPI or recombinant human TFPI-2 is observed. Reconstitute at 0.5 mg/mL in sterile PBS.

TFPI2 may play a role in the regulation of plasmin-mediated matrix remodeling. It inhibits trypsin, plasmin, factor VIIa/tissue factor and weakly factor Xa. TFPI2 has no effect on thrombin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TFPI2
Applications Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 375717
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 5% trehalose and No Preservative
Gene TFPI2
Gene Accession No. O35536
Gene Alias AV000670; Placental protein 5; PP5; PP5/TFPI-2; PP5FLJ21164; REF1; REF-1; retinal pigment epithelium cell factor 1; TFPI2; TFPI-2; TFPI-2REF1; Tissue factor pathway inhibitor 2
Gene Symbols TFPI2
Host Species Rat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse TFPI-2 Leu23-Lys211.
Purification Method Protein A/G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 21789
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.