Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TFPI2 Polyclonal Antibody
Goat Polyclonal Antibody
Supplier: Invitrogen™ PA547566
Description
In direct ELISAs and Western blots, less than 5% cross-reactivity with recombinant human (rh) TFPI-2, rhTFPI-1, and recombinant mouse TFPI-1 is observed. Reconstitute at 0.2 mg/mL in sterile PBS.
TFPI2 may play a role in the regulation of plasmin-mediated matrix remodeling. It inhibits trypsin, plasmin, factor VIIa/tissue factor and weakly factor Xa. TFPI2 has no effect on thrombin.
Specifications
TFPI2 | |
Polyclonal | |
Unconjugated | |
TFPI2 | |
AV000670; Placental protein 5; PP5; PP5/TFPI-2; PP5FLJ21164; REF1; REF-1; retinal pigment epithelium cell factor 1; TFPI2; TFPI-2; TFPI-2REF1; Tissue factor pathway inhibitor 2 | |
Goat | |
Antigen Affinity Chromatography | |
RUO | |
21789 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Immunoprecipitation, Western Blot | |
0.2 mg/mL | |
PBS with 5% trehalose and No Preservative | |
O35536 | |
TFPI2 | |
Mouse myeloma cell line NS0-derived recombinant mouse TFPI-2 Leu23-Lys211. | |
100 μg | |
Primary | |
Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction