Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TFPI2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA544873
Description
Peptide sequence: NANNFYTWEA CDDACWRIEK VPKVCRLQVS VDDQCEGSTE KYFFNLSSMT Sequence homology: Human: 100%.
TFPI2 may play a role in the regulation of plasmin-mediated matrix remodeling. It inhibits trypsin, plasmin, factor VIIa/tissue factor and weakly factor Xa. TFPI2 has no effect on thrombin.
Specifications
TFPI2 | |
Polyclonal | |
Unconjugated | |
TFPI2 | |
AV000670; Placental protein 5; PP5; PP5/TFPI-2; PP5FLJ21164; REF1; REF-1; retinal pigment epithelium cell factor 1; TFPI2; TFPI-2; TFPI-2REF1; Tissue factor pathway inhibitor 2 | |
Rabbit | |
Affinity chromatography | |
RUO | |
7980 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot, Immunocytochemistry | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
P48307 | |
TFPI2 | |
Synthetic peptide directed towards the middle region of human TFPI2. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction