Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TFPT Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA566177
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA566177 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA566177 Supplier Invitrogen™ Supplier No. PA566177
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PSGRKRRRVPR DGRRAGNALT PELAPVQIKV EEDFGFEADE ALDSSWVSRG PDKLLPYPTL Highest antigen sequence identity to the following orthologs - mouse 85%, rat 87%.

TFPT component of the chromatin-remodeling INO80 complex which, in addition to chromatin remodeling implicated in crucial functions such as DNA repair, checkpoint regulation, DNA replication, telomemere maintenance and chromosome segregation. This protein appears to promote apoptosis in a TP53/p53-independent manner.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TFPT
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene TFPT
Gene Accession No. P0C1Z6
Gene Alias 2400004F01Rik; AI450389; amida; amida, partner of the E2A; FB1; INO80 complex subunit F; INO80F; Protein amida; Protein FB1; TCF3 (E2A) fusion partner; TCF3 (E2A) fusion partner (in childhood Leukemia); TCF3 fusion partner; TCF3 fusion partner homolog; Tfpt
Gene Symbols TFPT
Host Species Rabbit
Immunogen Recombinant Human TFPT. Recombinant protein control fragment (Product #RP-106865).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 29844
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.