Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TfR (Transferrin R) Antibody (040), mFluor Violet 500 SE, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP289744MFV500
Description
TfR (Transferrin R) Monoclonal antibody specifically detects TfR (Transferrin R) in Human samples. It is validated for Flow CytometrySpecifications
TfR (Transferrin R) | |
Monoclonal | |
mFluor Violet 500 SE | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Flow Cytometry | |
40 | |
Flow Cytometry | |
CD71 antigen, CD71TRFR, p90, T9, TfR, TFR1, TR, transferrin receptor (p90, CD71), transferrin receptor protein 1, Trfr | |
This antibody was obtained from a rabbit immunized with purified, recombinant Human TfR (Transferrin R) (NP_003225.2; Cys89-Phe760). | |
0.1 mL | |
Cell Biology, Cellular Markers, Hematopoietic Stem Cell Markers, Immunology, Signal Transduction, Stem Cell Markers | |
7037 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction