Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TfR (Transferrin R) Antibody (8D3), Alexa Fluor™ 647, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals NB10064979AF647
Description
TfR (Transferrin R) Monoclonal specifically detects TfR (Transferrin R) in Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen.Specifications
TfR (Transferrin R) | |
Monoclonal | |
Alexa Fluor 647 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
TFRC | |
Mouse transformed endothelioma cell line t.end1. | |
0.25 mL | |
Cell Biology, Cellular Markers, Hematopoietic Stem Cell Markers, Immunology, Signal Transduction, Stem Cell Markers | |
7037 | |
Mouse | |
Purified |
Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen) | |
8D3 | |
Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen | |
CD71 antigen, CD71TRFR, p90, T9, TfR, TFR1, TR, transferrin receptor (p90, CD71), transferrin receptor protein 1, Trfr | |
Rat | |
Protein G purified | |
RUO | |
Primary | |
NB100-64979 recognizes native, soluble and denatured forms of murine CD71. The antibody has been used as a BBB transporter vector in mice and is suitable for studying CD71 expression, and iron uptake into different tissues, in the mouse. | |
Store at 4C in the dark. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction