Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TfR (Transferrin R) Antibody (OX26), DyLight 755, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB10079Z
This item is not returnable.
View return policy
Description
TfR (Transferrin R) Monoclonal antibody specifically detects TfR (Transferrin R) in Rat samples. It is validated for Flow CytometrySpecifications
TfR (Transferrin R) | |
Monoclonal | |
DyLight 755 | |
CD71 antigen, CD71TRFR, p90, T9, TfR, TFR1, TR, transferrin receptor (p90, CD71), transferrin receptor protein 1, Trfr | |
PHA activated rat lymphocytes | |
0.1 mL | |
Cell Biology, Cellular Markers, Hematopoietic Stem Cell Markers, Immunology, Signal Transduction, Stem Cell Markers | |
7037 | |
Store at 4°C in the dark. | |
IgG2a |
Flow Cytometry | |
OX26 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Primary | |
Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction