Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TfR (Transferrin R) Antibody (TFRC/1149), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP2450080.2MG
Description
Ensure accurate, reproducible results in Western Blot, Flow Cytometry, Immunofluorescence
TfR (Transferrin R) Monoclonal antibody specifically detects TfR (Transferrin R) in Human samples. It is validated for Western Blot, Flow Cytometry, Immunofluorescence.Specifications
TfR (Transferrin R) | |
Monoclonal | |
0.2 mg/mL | |
Western Blot 0.5 - 1.0 ug/ml, Flow Cytometry 0.5 - 1 ug/million cells in 0.1 ml, Immunofluorescence 1 - 2 ug/ml | |
CD71 antigen, CD71TRFR, p90, T9, TfR, TFR1, TR, transferrin receptor (p90, CD71), transferrin receptor protein 1, Trfr | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
It recognizes a ∼90-95kDa protein which is identified as cell surface transferrin receptor (CD71), a disulfide-bonded homodimeric glycoprotein of 180-190kDa. This MAb is highly specific to CD71 and shows no cross-reaction with other related proteins. Ligand for transferrin receptor is the serum iron transport protein, transferrin. This receptor is broadly distributed in carcinomas, sarcomas, leukemias, and lymphomas. CD71/Transferrin receptor has been reported to be associated with cell proliferation in both normal and neoplastic tissues and useful in predicting clinical behavior or response to therapy in a number of malignancies including breast cancer. | |
Store at 4C. | |
IgG1 κ |
Western Blot, Flow Cytometry, Immunofluorescence | |
TFRC/1149 | |
Unconjugated | |
10mM PBS and 0.05% BSA with 0.05% Sodium Azide | |
TFRC | |
Recombinant human TFRC protein | |
0.2 mg | |
Cell Biology, Cellular Markers, Hematopoietic Stem Cell Markers, Immunology, Signal Transduction, Stem Cell Markers | |
7037 | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction