Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TfR (Transferrin R) Antibody (YTA74.4) - BSA Free
SDP

Rat Monoclonal Antibody

$182.00 - $464.00

Specifications

Antigen TfR (Transferrin R)
Clone YTA74.4
Concentration 1.0 mg/mL
Dilution Flow Cytometry 1:10-1:1000, Immunohistochemistry 1:10-1:500, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500
Applications Flow Cytometry, Immunoprecipitation, Immunohistochemistry (Frozen), Immunohistochemistry
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB017719
SDP
View Documents
Novus Biologicals
NB100653970.025MG
0.025 mg
Each for $182.00
Only null left
Add to Cart
 
NB10065397
SDP
View Documents
Novus Biologicals
NB10065397
0.125 mg
Each for $464.00
Only null left
Add to Cart
 
Description

Description

TfR (Transferrin R) Monoclonal specifically detects TfR (Transferrin R) in Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Frozen.
Specifications

Specifications

TfR (Transferrin R)
1.0 mg/mL
Flow Cytometry, Immunoprecipitation, Immunohistochemistry (Frozen), Immunohistochemistry
Unconjugated
Rat
Cell Biology, Cellular Markers, Hematopoietic Stem Cell Markers, Immunology, Signal Transduction, Stem Cell Markers
Q62351
7037
Mouse Concanavilin A activated spleen cells.
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
YTA74.4
Flow Cytometry 1:10-1:1000, Immunohistochemistry 1:10-1:500, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500
Monoclonal
Purified
RUO
Mouse
CD71 antigen, CD71TRFR, p90, T9, TfR, TFR1, TR, transferrin receptor (p90, CD71), transferrin receptor protein 1, Trfr
TFRC
IgG2a
Protein G purified
Mouse CD71 antigen, the transferrin receptor (TFR). Expressed on all dividing cells and can distinguish resting and activated T-cells. Immunoprecipitates a disulphide linked homodimer of 90kD by SDS PAGE. The antibody inhibits cell proliferation and the mixed lymphocyte response in vitro. Blocks the binding of R17 217.1.3. and R17 208.2 anti-TFR monoclonal antibodies (1,2).
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.