Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TfR (Transferrin R) Antibody (040), Alexa Fluor™ 488, Novus Biologicals™
SDP

Catalog No. NB109082 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB109082 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB109082 Supplier Novus Biologicals Supplier No. NBP289744AF488
Only null left
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

TfR (Transferrin R) Monoclonal specifically detects TfR (Transferrin R) in Human samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen TfR (Transferrin R)
Applications Flow Cytometry, Immunofluorescence
Classification Monoclonal
Clone 40
Conjugate Alexa Fluor 488
Formulation 50 mM Sodium Borate
Gene Alias CD71 antigen, CD71TRFR, p90, T9, TfR, TFR1, TR, transferrin receptor (p90, CD71), transferrin receptor protein 1, Trfr
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Human TfR (Transferrin R) (NP_003225.2; Cys89-Phe760).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Biology, Cellular Markers, Hematopoietic Stem Cell Markers, Immunology, Signal Transduction, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 7037
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.