Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TfR (Transferrin R) Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

$243.50

Specifications

Antigen TfR (Transferrin R)
Applications Western Blot, Immunohistochemistry, KnockDown
Classification Polyclonal
Conjugate Unconjugated
Form Purified
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB092958
SDP
View Documents
Novus Biologicals
NBP30327120UL
20 μL
Each for $243.50
Only null left
Add to Cart
 
NB092957
SDP
View Documents
Novus Biologicals
NBP303271100UL
100 μL N/A N/A N/A
Description

Description

TfR (Transferrin R) Polyclonal specifically detects TfR (Transferrin R) in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Knockout Validated.
Specifications

Specifications

TfR (Transferrin R)
Polyclonal
Purified
RUO
Human, Mouse, Rat
CD71 antigen, CD71TRFR, p90, T9, TfR, TFR1, TR, transferrin receptor (p90, CD71), transferrin receptor protein 1, Trfr
Recombinant fusion protein containing a sequence corresponding to amino acids 1-140 of human TfR (Transferrin R) (NP_001121620.1).
Primary
Store at -20°C. Avoid freeze-thaw cycles.
Western Blot, Immunohistochemistry, KnockDown
Unconjugated
Rabbit
Cell Biology, Cellular Markers, Hematopoietic Stem Cell Markers, Immunology, Signal Transduction, Stem Cell Markers
PBS with 50% glycerol, pH7.3.
7037
IgG
Affinity purified
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.