Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TfR1 Mouse anti-Human, Clone: 3B82A1, MilliporeSigma™

Catalog No. MABS1982MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABS1982MI 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABS1982MI Supplier MilliporeSigma Supplier No. MABS1982
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Detects Transferrin receptor protein 1 using this mouse monoclonal Anti-TfR1, clone 3B82A1 Immunocytochemistry, Immunoprecipitation, and Western Blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TfR1
Applications Flow Cytometry, Western Blot, Immunocytochemistry, Immunoprecipitation
Classification Monoclonal
Clone 3B82A1
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Gene Symbols TFRC
Host Species Mouse
Immunogen His-tagged recombinant ectodomain of human Transferrin receptor protein 1.
Purification Method Protein G Purified
Quantity 200 μL
Regulatory Status RUO
Research Discipline Signaling
Primary or Secondary Primary
Gene ID (Entrez) NP_001121620.1
Test Specificity Clone 3B82A1 specifically detects transferrin receptor protein 1. It targets an epitope in the extracellullar domain.
Target Species Human
Content And Storage Stable for 1 year at 2-8°C from date of receipt.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.