Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TfR2 Mouse anti-Human, Clone: 9F8 1C11, MilliporeSigma™

Catalog No. MABS1981MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABS1981MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABS1981MI Supplier MilliporeSigma Supplier No. MABS1981
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-TfR2, clone 9F8-1C11 Antibody is a highly specific mouse monoclonal antibody, that targets Tfr2 and has been tested in Western Blotting, Immunoprecipitation, and Immunocytochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TfR2
Applications Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 9F8 1C11
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Gene Symbols TRF2
Host Species Mouse
Immunogen Purified ectodomains of the human TfR2.
Purification Method Protein G Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Epigenetics and Nuclear Function
Primary or Secondary Primary
Gene ID (Entrez) NP_003218.2
Test Specificity Clone 9F8-1C11 sprcifically detects human TfR2 and does exhibit reactivity with TfR.
Target Species Human
Content And Storage Stable for 1 year at 2-8°C from date of receipt.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.