Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TFR2 Mouse anti-Human, Mouse, Clone: OTI1B1, liquid, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene TA504592

Description
This gene is a member of the transferrin receptor-like family and encodes a single-pass type II membrane protein with a protease associated (PA) domain, an M28 peptidase domain and a transferrin receptor-like dimerization domain. This protein mediates cellular uptake of transferrin-bound iron and mutations in this gene have been associated with hereditary hemochromatosis type III. Alternatively spliced variants which encode different protein isoforms have been described; however, not all variants have been fully characterized.Specifications
TFR2 | |
Monoclonal | |
1.13 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
Q9JKX3, Q9UP52 | |
TFR2 | |
Full length human recombit protein of human TFR2 produced in HEK293T cell. | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG2a |
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Western Blot | |
OTI1B1 | |
Unconjugated | |
TFR2 | |
HFE3, TFRC2 | |
Mouse | |
Affinity Chromatography | |
RUO | |
50765, 7036 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction