Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TFR2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA529137
Description
Recommended positive controls: K562, HL-60. Predicted reactivity: Mouse (89%), Rat (88%), Bovine (89%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
This gene is a member of the transferrin receptor-like family and encodes a single-pass type II membrane protein with a protease associated (PA) domain, an M28 peptidase domain and a transferrin receptor-like dimerization domain. This protein mediates cellular uptake of transferrin-bound iron and mutations in this gene have been associated with hereditary hemochromatosis type III. Alternatively spliced variants which encode different protein isoforms have been described; however, not all variants have been fully characterized.
Specifications
| TFR2 | |
| Polyclonal | |
| Unconjugated | |
| Tfr2 | |
| HFE3; MGC126368; TfR2; TFRC2; transferrin receptor 2; transferrin receptor protein 2; Trfr2 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 7036 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 | |
| Q9UP52 | |
| Tfr2 | |
| Recombinant protein encompassing a sequence within the Extracellular domain of human TFR2. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction