Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Abnova™ TFRC (Human) ELISA Kit
Click to view available options
Quantity:
1 Kit
Description
TFRC (Human) ELISA Kit is a sandwich enzyme-linked immunosorbent assay for the quantitative measurement of human TFRC in serum, plasma, tissue homogenates, culture supernatants and other biological fluids.
Specifications
Specifications
Product Type | ELISA Kit |
Target Species | Human |
For Use With (Application) | Quant |
Gene ID (Entrez) | 7037 |
Quantity | 1 Kit |
Storage Requirements | Store the kit at -20°C |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction