Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TFRC Mouse anti-Human, Clone: 236-15375, Abnova™

Catalog No. 89035726 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-035-726 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-035-726 Supplier Abnova Corporation Supplier No. MAB14185

Mouse Monoclonal Antibody

Mouse monoclonal antibody raised against human TFRC.

  • High specificity
  • High reactivity

Specifications

Antigen TFRC
Applications Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone 236-15375
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against human TFRC.
Dilution Immunocytochemistry (1 ug/mL) Immunofluorescence The optimal working dilution should be determined by the end user.
Formulation Lyophilized.
Gene Accession No. P02786
Gene Alias CD71/TFR/TFR1/TRFR
Gene Symbols TFRC
Host Species Mouse
Immunogen Human TFRC.
Purification Method Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7037
Target Species Human
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Lyophilized
Isotype IgG1
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.