Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TFRC Mouse anti-Human, Clone: OTI6H9, lyophilized, TrueMAB™
SDP

Catalog No. 501666607
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-166-6607 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-166-6607 Supplier OriGene Supplier No. CF803640
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO

Transferrin Receptor (CD71) is a type II transmembrane disulfide-bonded glycoprotein expressed as homodimer in erythroid blood cell line and activated leukocytes. The ligand for Transferrin Receptor is the serum iron transport protein, transferrin. Upon binding holotransferrin (complex of transferrin and iron ions), Transferrin Receptor is internalized by clathrin-mediated endocytosis. Acidification of endosomes by vesicular membrane proton pumps leads to dissociation of iron ions, whereas transferrin (apotransferrin) remains associated with Transferrin Receptor and recycles to the cell surface, where it is released upon exposure to neutral pH and the concomitant loss of affinity of apotransferrin for its receptor. Transferrin Receptor is also involved in uptake of non-transferrin bound iron. Transferrin Receptor is necessary for development of erythrocytes and the nervous system. Serum Transferrin Receptor (sTfR) is used as a means of detecting erythropoietin (EPO) misuse by athletes and as a diagnostic test for anemias resulting from conditions including rheumatoid arthritis, pregcy, irritable bowel syndrome and HIV infection. Transferrin Receptor has been reported to be associated with cell proliferation in both normal and neoplastic tissues and useful in predicting clinical behavior or response to therapy in maligcies such as breast cancer.

Specifications

Antigen TFRC
Applications Western Blot
Classification Monoclonal
Clone OTI6H9
Conjugate Unconjugated
Formulation PBS with 8% trehalose and no preservative; pH 7.3
Gene TFRC
Gene Accession No. P02786
Gene Alias CD71, IMD46, T9, TFR, TFR1, TR, TRFR, p90
Gene Symbols TFRC
Host Species Mouse
Immunogen Full length human recombit protein of human TFRC produced in HEK293T cell.
Purification Method Affinity Chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7037
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.