Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TGDS Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5112739
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5112739 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5112739 Supplier Invitrogen™ Supplier No. PA5112739
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

TGDS Polyclonal Antibody for Western Blot, IHC (P), ELISA

TRUSTED_SUSTAINABILITY

Specifications

Antigen TGDS
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 3 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene TGDS
Gene Accession No. O95455
Gene Alias 2610017J16Rik; 2610025M23Rik; AI648925; CATMANS; dTDP-D-glucose 4,6-dehydratase; growth-inhibiting protein 21; SDR2E1; short chain dehydrogenase/reductase family 2E, member 1; TDPGD; TDP-glucose 4,6-dehydratase; TGDS
Gene Symbols TGDS
Host Species Rabbit
Immunogen Recombinant Human dTDP-D-glucose 4,6-dehydratase protein (143-305AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23483, 306164
Target Species Human, Rat
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.