Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TGF-alpha Antibody (P/T1) - Azide and BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP2347020.1MG
Description
TGF-alpha Monoclonal antibody specifically detects TGF-alpha in Human, Rabbit, Zebrafish samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin), CyTOFSpecifications
TGF-alpha | |
Monoclonal | |
1.0 mg/mL | |
Flow Cytometry 0.5 to 1 μg/million cells, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence 1 to 2 μg/mL, Immunohistochemistry-Paraffin 1 to 2 μg/mL, CyTOF-ready | |
protransforming growth factor alpha, TFGA, transforming growth factor, alpha, transforming growth factor-alpha | |
A 10-amino acid synthetic peptide (aa34-43; PPVAAAVVSH) from human TGF-alpha (Uniprot: P01135) | |
0.1 mg | |
Angiogenesis, Cell Cycle and Replication, Signal Transduction | |
7039 | |
Store at -20°C to -80°C. Avoid freeze-thaw cycles. | |
IgG1 κ |
Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin), CyTOF | |
P/T1 | |
Unconjugated | |
10 mM PBS | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human, Rabbit, Zebrafish | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction