Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TGF-alpha Antibody (tAb2), mFluor Violet 610 SE, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP281082MFV610
Description
TGF-alpha Monoclonal antibody specifically detects TGF-alpha in Human samples. It is validated for Flow CytometrySpecifications
TGF-alpha | |
Monoclonal | |
mFluor Violet 610 SE | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Flow Cytometry | |
tAb2 | |
Flow Cytometry | |
protransforming growth factor alpha, TFGA, transforming growth factor, alpha, transforming growth factor-alpha | |
Biologically active hemocyanin-coupled human TGF-alpha. | |
0.1 mL | |
Angiogenesis, Cell Cycle and Replication, Signal Transduction | |
7039 | |
Store at 4C in the dark. | |
IgG κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction