Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TGF-alpha Antibody (TGFA/1119), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP2451420.1MG
Description
Ensure accurate, reproducible results in Flow Cytometry, Immunohistochemistry (Paraffin), Immunofluorescence
TGF-alpha Monoclonal specifically detects TGF-alpha in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
TGF-alpha | |
Monoclonal | |
0.2mg/mL | |
Flow Cytometry 0.5 - 1 ug/million cells in 0.1 ml, Immunohistochemistry-Paraffin 2 - 4ug/ml, Immunofluorescence 1 - 2 ug/ml | |
P01135 | |
TGFA | |
Recombinant human full-length TGF alpha protein | |
0.1 mg | |
Angiogenesis, Cell Cycle and Replication, Signal Transduction | |
7039 | |
Human | |
Purified |
Flow Cytometry, Immunohistochemistry (Paraffin), Immunofluorescence | |
TGFA/1119 | |
Unconjugated | |
10mM PBS and 0.05% BSA with 0.05% Sodium Azide | |
protransforming growth factor alpha, TFGA, transforming growth factor, alpha, transforming growth factor-alpha | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
This antibody reacts with the C-terminus of TGF alpha and shows no cross-reaction with EGF and the neuropeptide synenkephalin. TGF alpha (aa50) is a growth factor with 33% homology to EGF, binds to EGFR, activates tyrosine phosphorylation of the receptor, and stimulates cell proliferation. It plays a role in tumor initiation by inducing the reversible transformed phenotype. | |
Store at 4C. | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction