Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TGF alpha Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA586743
Description
Purity is > 95% by SDS-PAGE.
TGF-alpha is an EGF-related polypeptide growth factor that signals through the EGF receptor, and stimulates the proliferation of a wide range of epidermal and epithelial cells. It is produced by monocytes, keratinocytes, and various tumor cells. TGF-alpha induces anchorage-independence transformation in cultured cells. Human, murine and rat TGF-alpha are cross-species reactive. TGFalpha (aa 50) is a growth factor with 33% homology to EGF, binds to EGFR, activates tyrosine phosphorylation of the receptor, and stimulates cell proliferation. It plays a role in tumor initiation by inducing the reversible transformed phenotype.
Specifications
TGF alpha | |
Polyclonal | |
Unconjugated | |
TGFA | |
EGF-like TGF; ETGF; I79_019067; LOC100009150; Protransforming growth factor alpha; RATTGFAA; TFGA; TGF a; TGF type 1; TGF a; TGFA; TGFAA; TGFalpha; TGF-alpha; TGFa; Transforming growth factor; Transforming growth factor alpha; transforming growth factor, alpha; transforming growth factor-alpha; wa1; wa-1; waved 1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
21802, 24827, 7039 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
P01134, P01135, P48030 | |
TGFA | |
A synthetic peptide corresponding to residues in Human TGFalpha. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction