Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TGF alpha Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5120044
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5120044 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5120044 Supplier Invitrogen™ Supplier No. PA5120044
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Positive test controls include: 293T, Jurkat, Mouse lung. The target is usually found in the following locations: Cell membrane, Secreted, Single-pass type I membrane protein, extracellular space. Immunogen sequence: ENSTSPLSDP PVAAAVVSHF NDCPDSHTQF CFHGTCRFLV QEDKPACVCH SGYVGARCEH ADLLAVVAAS QKKQA.

TGF-alpha is an EGF-related polypeptide growth factor that signals through the EGF receptor, and stimulates the proliferation of a wide range of epidermal and epithelial cells. It is produced by monocytes, keratinocytes, and various tumor cells. TGF-alpha induces anchorage-independence transformation in cultured cells. Human, murine and rat TGF-alpha are cross-species reactive. TGFalpha (aa 50) is a growth factor with 33% homology to EGF, binds to EGFR, activates tyrosine phosphorylation of the receptor, and stimulates cell proliferation. It plays a role in tumor initiation by inducing the reversible transformed phenotype.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TGF alpha
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.93 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene TGFA
Gene Accession No. P01135, P48030
Gene Alias EGF-like TGF; ETGF; I79_019067; LOC100009150; Protransforming growth factor alpha; RATTGFAA; TFGA; TGF a; TGF type 1; TGF α; TGFA; TGFAA; TGFalpha; TGF-alpha; TGFα; Transforming growth factor; Transforming growth factor alpha; transforming growth factor, alpha; transforming growth factor-alpha; wa1; wa-1; waved 1
Gene Symbols TGFA
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 24-98 of human TGFA (NP_001093161.1).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 21802, 7039
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.