Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TGF-alpha Polyclonal Antibody, Abbomax

Catalog No. 502203714
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-220-3714 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-220-3714 Supplier Abbomax Inc Supplier No. 5003734
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive control: Human liver and hepatocellular carcinoma tissues. Cellular location: Cell membrane and Secreted.

TGF-alpha Polyclonal antibody specifically detects TGF-alpha in Human samples. It is validated for ELISA, Immunohistochemistry (Paraffin)

Specifications

Antigen TGF-alpha
Applications ELISA, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with proprietary stabilizer and 0.01% sodium azide
Gene TGFA
Gene Accession No. P01135
Gene Alias EGF-like TGF; ETGF; I79_019067; LOC100009150; Protransforming growth factor alpha; RATTGFAA; TFGA; TGF a; TGF type 1; TGF a; TGFA; TGFAA; TGFalpha; TGF-alpha; TGFa; Transforming growth factor; Transforming growth factor alpha; transforming growth factor, alpha; transforming growth factor-alpha; wa1; wa-1; waved 1
Host Species Rabbit
Immunogen A synthetic peptide corrsponding to the C-terminus of human TGF-alpha
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7039
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.