Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TGF-beta 1, 2, 3 APC-conjugated Antibody, R&D Systems™
Mouse Monoclonal Antibody has been used in 2 publications
Supplier: R&D Systems IC1835A
This item is not returnable.
View return policy
Description
TGF beta 1/2/3 Monoclonal specifically detects TGF beta 1/2/3 in Multi-Species samples. It is validated for Intracellular Staining by Flow Cytometry.Specifications
TGF beta 1/2/3 | |
Monoclonal | |
APC | |
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
TGFB1 | |
Bovine bone-derived TGF-beta 1 and TGF-beta 2 | |
100 Tests | |
Primary | |
Detects bovine, chicken, mouse, and human TGF-beta in ELISAs and Western blots. It recognizes human TGF-beta 1, TGF-beta 2, and TGF-beta 3. | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
IgG1 |
Flow Cytometry | |
1D11 | |
Intracellular Staining by Flow Cytometry 10 uL/10^6 cells | |
ARVD1, CED, DPD1, LAP, LDS5, RNHF, TGF beta1, TGFB, TGFB1, TGFB2, TGFB3, TGFbeta, TGFbeta 1, TGFbeta 2, TGFbeta 3, transforming growth factor, beta 1 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
7040 | |
Multi-species | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction