Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TGF-beta 1, 2, 3 PE-conjugated Antibody, R&D Systems™

Catalog No. IC1835P
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
IC1835P 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. IC1835P Supplier R&D Systems Supplier No. IC1835P

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

This item is not returnable. View return policy

Mouse Monoclonal Antibody has been used in 2 publications

TGF beta 1/2/3 Monoclonal specifically detects TGF beta 1/2/3 in Multi-Species samples. It is validated for Intracellular Staining by Flow Cytometry.

Specifications

Antigen TGF beta 1/2/3
Applications Flow Cytometry
Classification Monoclonal
Clone 1D11
Conjugate R-PE
Dilution Intracellular Staining by Flow Cytometry 10 uL/10^6 cells
Formulation Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
Gene Alias ARVD1, CED, DPD1, LAP, LDS5, RNHF, TGF beta1, TGFB, TGFB1, TGFB2, TGFB3, TGFbeta, TGFbeta 1, TGFbeta 2, TGFbeta 3, transforming growth factor, beta 1
Gene Symbols TGFB1
Host Species Mouse
Immunogen Bovine bone-derived TGF-beta 1 and TGF-beta 2
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7040
Test Specificity Detects bovine, chicken, mouse, and human TGF-beta in ELISAs and Western blots. It recognizes human TGF-beta 1, TGF-beta 2, and TGF-beta 3.
Target Species Multi-species
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.