Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TGF-beta 1 Antibody (016), DyLight 488, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP289646G
Description
TGF-beta 1 Monoclonal antibody specifically detects TGF-beta 1 in Human samples. It is validated for ELISASpecifications
TGF-beta 1 | |
Monoclonal | |
DyLight 488 | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
ELISA | |
016 | |
ELISA | |
CEDLAP, DPD1, latency-associated peptide, TGF beta, TGFB, TGFbeta, TGF-beta 1 protein, TGF-beta-1, transforming growth factor beta-1, transforming growth factor, beta 1 | |
This antibody was obtained from a rabbit immunized with purified, recombinant Human TGF-beta 1 (NP_000651.3; Met 1-Ser 390). | |
0.1 mL | |
Angiogenesis, Apoptosis, Asthma, Cancer, Immunology, Phospho Specific, Signal Transduction | |
7040 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction