Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TGF-beta 1 Antibody (MM0446-6G14), Alexa Fluor™ 350, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP211766AF350
Description
TGF-beta 1 Monoclonal specifically detects TGF-beta 1 in Human samples. It is validated for Flow Cytometry.Specifications
TGF-beta 1 | |
Monoclonal | |
Alexa Fluor 350 | |
50 mM sodium borate with 0.05% sodium azide | |
TGFB1 | |
Human recombinant TGF-beta 1 | |
0.1 mL | |
Primary | |
Human | |
IgG1 |
Western Blot, Flow Cytometry | |
MM0446-6G14 | |
Western Blot, Flow Cytometry | |
CEDLAP, DPD1, latency-associated peptide, TGFB, TGFbeta, TGF-beta 1 protein, TGF-beta-1, transforming growth factor beta-1, transforming growth factor, beta 1 | |
Mouse | |
Protein G purified | |
Angiogenesis, Apoptosis, Asthma, Cancer, Immunology, Phospho Specific, Signal Transduction | |
7040.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction