Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TGF-beta 1 Antibody (MM0446-6G14) - Azide and BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
$206.50 - $560.50
Specifications
| Antigen | TGF-beta 1 |
|---|---|
| Clone | MM0446-6G14 |
| Concentration | LYOPH |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
Description
TGF-beta 1 Monoclonal specifically detects TGF-beta 1 in Human samples. It is validated for Flow Cytometry, CyTOF-ready, Block/Neutralize.Specifications
| TGF-beta 1 | |
| LYOPH | |
| Unconjugated | |
| Mouse | |
| Angiogenesis, Apoptosis, Asthma, Cancer, Immunology | |
| P01137 | |
| 7040 | |
| IgG1 | |
| Protein G purified |
| MM0446-6G14 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| CEDLAP, DPD1, latency-associated peptide, TGFB, TGFbeta, TGF-beta 1 protein, TGF-beta-1, transforming growth factor beta-1, transforming growth factor, beta 1 | |
| TGFB1 | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title