Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PeproTech TGF beta-1 Monoclonal Antibody, PeproTech®

Catalog No. 500M66500UG
Encompass_Preferred
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
500M66500UG 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 500M66500UG Supplier PeproTech Supplier No. 500M66500UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

AA Sequence of recombinant protein: discontinued. Preparation: Produced in BALB/c mice immunized with highly pure Recombinant Human TGF-beta1. Anti-Human TGF-beta1-specific antibody was purified from tissue culture supernatant by Protein G affinity chromatography. Western Blot: To detect Human TGF-beta1 by Western Blot analysis this antibody can be used at a concentration of 2.0-4.0 μg/mL. When used in conjunction with compatible secondary reagents the detection limit for Recombinant Human TGF-beta1 is 0.25-0.50 ng/lane, under reducing and non-reducing conditions.

TGF beta-1 (TGFB1, Transforming Growth Factor Beta 1) is a polypeptide member of the transforming growth factor beta superfamily of cytokines, found almost ubiquitously in tissues. Transforming growth factor (TGF)-b is stored in the extracellular matrix as a latent complex with its pro-domain. Activation of TGF beta-1 requires the binding of aV integrin to an RGD sequence in the prodomain and exertion of force on this domain, which is held in the extracellular matrix by latent TGF-b binding proteins. Latent forms are complexes of TGF beta-1, an amino-terminal portion of the TGF-beta precursor, designated TGF-LAP (TGF-latency associated peptide), and a specific binding protein, known as LTBP. TGF beta-1 helps regulates proliferation, differentiation, adhesion, migration in many cell types. Many cells have TGF beta receptors, and the protein positively and negatively regulates many other growth factors. TGF beta-1 is cleaved into a latency-associated peptide and a mature TGF beta-1 peptide, and is found in either a latent form composed of a TGFB1 homodimer, a LAP homodimer, and a latent TGFB1-binding protein, or in an active form composed of a TGF beta-1 homodimer. The mature peptide may also form heterodimers with other TGF beta family members. The gene for TGF beta-1 is frequently upregulated in tumor cells, and mutations in this gene result in Camurati-Engelmann disease and cystic fibrosis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TGF beta-1
Applications Western Blot
Classification Monoclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene Tgfb1
Gene Accession No. P01137
Gene Alias CED; cytokine; DPD1; LAP; Latency-associated peptide; prepro-transforming growth factor beta-1; regulatory protein; TGF b; TGF b 1; TGF beta; tgf beta 1; tgf beta1; TGF beta-1; TGF beta-1 protein; TGF β; TGF β 1; TGF β1; Tgfb; TGFB1; TGF-B1; Tgfb-1; TGFbeta; TGF-beta 1; TGF-beta 1 protein; TGFbeta1; TGF-beta1; TGF-beta-1; TGFβ1; Transforming growth factor; transforming growth factor b1; transforming growth factor beta; transforming growth factor beta 1; transforming growth factor beta-1; Transforming growth factor beta-1 proprotein; transforming growth factor beta-1 proprotein; transforming growth factor beta-1; transforming growth factor β1; transforming growth factor, beta 1; transforming growth factor, beta 1 (Camurati-Engelmann disease); transforming growth factor, beta-1; transforming growth factor-beta 1; transforming growth factor-beta I; transforming growth factor-beta-1; transforming growth factor-beta-1 precursor; unnamed protein product
Gene Symbols Tgfb1
Host Species Mouse
Immunogen Insect cells-derived Recombinant Human TGF-beta1.
Purification Method Protein G
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7040
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Lyophilized
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.