Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TGF beta 1 Mouse anti-Human, Clone: 1E3E9, ProteinTech
SDP

Catalog No. 502239951 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-223-9951 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 50-223-9951 Supplier Proteintech Group Inc Supplier No. 690121IG100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Reconstitute with water to the desired concentration. Lyophilized antibodies are stable for 1 year from the date of purchase if stored between -20 to -80°C. Upon reconstitution, we recommend that the solution be stored at 4°C (short- term) or at -20 to -80°Cfor long term. Repeated freeze-thaw cycles should be avoided with reconstituted products. Aliquoting is unnecessary for -20°C storage.

TGF beta 1 Monoclonal antibody specifically detects TGF beta 1 in Human samples. It is validated for Flow Cytometry, Neutralization

Specifications

Antigen TGF beta 1
Applications Flow Cytometry, Neutralization
Classification Monoclonal
Clone 1E3E9
Conjugate Unconjugated
Formulation PBS with no preservative
Gene Tgfb1
Gene Accession No. P01137
Gene Alias CED; cytokine; DPD1; LAP; Latency-associated peptide; prepro-transforming growth factor beta-1; regulatory protein; TGF b; TGF b 1; TGF beta; tgf beta 1; tgf beta1; TGF beta-1; TGF beta-1 protein; TGF β; TGF β 1; TGF β1; Tgfb; TGFB1; TGF-B1; Tgfb-1; TGFbeta; TGF-beta 1; TGF-beta 1 protein; TGFbeta1; TGF-beta1; TGF-beta-1; TGFβ1; Transforming growth factor; transforming growth factor b1; transforming growth factor beta; transforming growth factor beta 1; transforming growth factor beta-1; Transforming growth factor beta-1 proprotein; transforming growth factor beta-1 proprotein; transforming growth factor beta-1; transforming growth factor β1; transforming growth factor, beta 1; transforming growth factor, beta 1 (Camurati-Engelmann disease); transforming growth factor, beta-1; transforming growth factor-beta 1; transforming growth factor-beta I; transforming growth factor-beta-1; transforming growth factor-beta-1 precursor; unnamed protein product
Gene Symbols Tgfb1
Host Species Mouse
Immunogen NeutraKine TGF beta 1 fusion protein HZ-1011
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7040
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Lyophilized
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.