Learn More
Invitrogen™ TGF beta-1 Recombinant Rabbit Monoclonal Antibody (4A4P3)
Rabbit Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA551415
Description
Immunogen sequence: LSTCKTIDME LVKRKRIEAI RGQILSKLRL ASPPSQGEVP PGPLPEAVLA LYNSTRDRVA GESAEPEPEP EADYYAKEVT RVLMVETHNE IYDKFKQSTH SIYMFFNTSE LREAVPEPVL LSRAELRLLR LKLKVEQHVE LYQKYSNNSW RYLSNRLLAP SDSPEWLSFD VTGVVRQWLS RGGEIEGFRL SAHCSCDSRD NTLQVDINGF TTGRRGDLAT IHGMNRPFLL LMATPLERAQ HLQSSRHRRA LDTNYCFSST EKNCCVRQLY IDFRKDLGWK WIHEPKGYHA NFCLGPCPYI WSLDTQYSKV LALYNQHNPG ASAAPCCVPQ ALEPLPIVYY VGRKPKVEQL SNMIVRSCKC S.
TGF beta-1 (TGFB1, Transforming Growth Factor Beta 1) is a polypeptide member of the transforming growth factor beta superfamily of cytokines, found almost ubiquitously in tissues. Transforming growth factor (TGF)-b is stored in the extracellular matrix as a latent complex with its pro-domain. Activation of TGF beta-1 requires the binding of aV integrin to an RGD sequence in the prodomain and exertion of force on this domain, which is held in the extracellular matrix by latent TGF-b binding proteins. Latent forms are complexes of TGF beta-1, an amino-terminal portion of the TGF-beta precursor, designated TGF-LAP (TGF-latency associated peptide), and a specific binding protein, known as LTBP. TGF beta-1 helps regulates proliferation, differentiation, adhesion, migration in many cell types. Many cells have TGF beta receptors, and the protein positively and negatively regulates many other growth factors. TGF beta-1 is cleaved into a latency-associated peptide and a mature TGF beta-1 peptide, and is found in either a latent form composed of a TGFB1 homodimer, a LAP homodimer, and a latent TGFB1-binding protein, or in an active form composed of a TGF beta-1 homodimer. The mature peptide may also form heterodimers with other TGF beta family members. The gene for TGF beta-1 is frequently upregulated in tumor cells, and mutations in this gene result in Camurati-Engelmann disease and cystic fibrosis.
Specifications
TGF beta-1 | |
Recombinant Monoclonal | |
0.6 mg/mL | |
PBS with 0.05% BSA, 50% glycerol and 0.05% ProClin 300; pH 7.3 | |
P01137, P17246 | |
Tgfb1 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 30-390 of human TGF beta 1 (NP_000651.3). | |
100 μL | |
Primary | |
Human, Rat | |
Antibody | |
Liquid |
ELISA, Western Blot, Immunocytochemistry | |
4A4P3 | |
Unconjugated | |
Tgfb1 | |
CED; cytokine; DPD1; LAP; Latency-associated peptide; prepro-transforming growth factor beta-1; regulatory protein; TGF b; TGF b 1; TGF beta; tgf beta 1; tgf beta1; TGF beta-1; TGF beta-1 protein; TGF ß; TGF ß 1; TGF ß1; Tgfb; TGFB1; TGF-B1; Tgfb-1; TGFbeta; TGF-beta 1; TGF-beta 1 protein; TGFbeta1; TGF-beta1; TGF-beta-1; TGFß1; Transforming growth factor; transforming growth factor b1; transforming growth factor beta; transforming growth factor beta 1; transforming growth factor beta-1; Transforming growth factor beta-1 proprotein; transforming growth factor beta-1 proprotein; transforming growth factor beta-1; transforming growth factor ß1; transforming growth factor, beta 1; transforming growth factor, beta 1 (Camurati-Engelmann disease); transforming growth factor, beta-1; transforming growth factor-beta 1; transforming growth factor-beta I; transforming growth factor-beta-1; transforming growth factor-beta-1 precursor; unnamed protein product | |
Rabbit | |
Affinity Chromatography | |
RUO | |
59086, 7040 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.