Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TGF-beta 2 Antibody (MM0640-6M28) - Azide and BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
$206.50
Specifications
Antigen | TGF-beta 2 |
---|---|
Clone | MM0640-6M28 |
Concentration | LYOPH |
Applications | Western Blot, Immunohistochemistry (Paraffin), Immunohistochemistry |
Classification | Monoclonal |
Description
TGF-beta 2 Monoclonal specifically detects TGF-beta 2 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
TGF-beta 2 | |
LYOPH | |
Monoclonal | |
Purified | |
RUO | |
Human | |
BSC-1 cell growth inhibitor, Cetermin, Glioblastoma-derived T-cell suppressor factor, G-TSF, MGC116892, Polyergin, TGF-beta2, TGF-beta-2, transforming growth factor beta-2, transforming growth factor, beta 2 | |
TGFB2 | |
Primary | |
Store at -20C. Avoid freeze-thaw cycles. |
MM0640-6M28 | |
Western Blot, Immunohistochemistry (Paraffin), Immunohistochemistry | |
Unconjugated | |
Mouse | |
Angiogenesis, Apoptosis, Biologically Active Proteins, Cell Cycle and Replication | |
P61812 | |
7042 | |
IgG2 | |
Protein G purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title